missing translation for 'onlineSavingsMsg'
Learn More

Lithium Carbonate, Extra Pure, SLR, Fisher Chemical

Product Code. 10244200 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic powder jar
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10244200 250 g Plastic powder jar 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10244200 Supplier Fisher Chemical Supplier No. L/2100/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifica

Melting Point 720°C
CAS 554-13-2
Color White
pH 10 to 11
Physical Form Powder/Solid
Quantity 250 g
Molecular Formula CLi2O3
MDL Number MFCD00011084
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
SMILES [Li+].[Li+].[O-]C([O-])=O
IUPAC Name dilithium(1+) carbonate
Molecular Weight (g/mol) 73.89
PubChem CID 11125
ChEBI CHEBI:6504
Formula Weight 73.88g/mol
Packaging Plastic powder jar
Vapor Pressure 1 hPa 610
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • Lithium carbonate
Avvertenza
  • Warning
Categoria di pericolo
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Indicazioni di pericolo
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P270-Do not eat, drink or smoke when using this product.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.