missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium carbonate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2 kg
10 kg
Description
Lithium carbonate-based glasses are used in overware. Used in the manufacture of lithium-ion battery cathodes. Pharmaceutical composition of lithium carbonate has been shown to exhibit antimanic effects specifically to treat mania associated with bipolar disorder. Used in both low-fire and high-fire ceramic glaze, tile adhesives, and in the processing of metal oxides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1310°C(decomposition) |
Melting Point | 723°C |
Color | White |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 99% |
Solubility Information | Soluble in water and dilute acids. Insoluble in alcohol. |
IUPAC Name | dilithium(1+) carbonate |
Formula Weight | 73.9 |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'