missing translation for 'onlineSavingsMsg'
Learn More

Lithium carbonate, Puratronic™, 99.998% (metals basis excluding Ca), Ca LT 20ppm

Product Code. p-4407560
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 11316817

Brand: Thermo Scientific Alfa Aesar 010734.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 554-13-2 | CLi2O3 | 73.89 g/mol

Stainless Steel powder is employed in pharmaceuticals, marine applications, architectural applications, medical implants, including pins, screws and orthopaedic implants like total hip and knee replacements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Boiling Point 1310°C (decomposition)
Melting Point 723°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.998% (Metals basis excluding Ca)
Solubility Information Soluble in water and acid. Insoluble in acetone,ammonia and alcohol.
IUPAC Name dilithium(1+) carbonate
Formula Weight 73.9
Odor Odorless
Appearance White powder
Density 2.11 g/cm3 at 20°C
Chemical Name or Material Lithium carbonate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.