missing translation for 'onlineSavingsMsg'
Learn More

Gratnells™ Stainless Steel Tray Trolley with Runners

Product Code. 13170173
Encompass
Click to view available options
Dimensions (L x W x H):
370 x 420 x 850 mm
710 x 420 x 850 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 13170173

Brand: Gratnells™ 2025GWR

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use Gratnells™ Stainless Steel Tray Trolley with Runners to stack and transport trays. Trolleys are supplied with runners.

  • 25mm square steel tubing finished in white epoxy coating with 75mm heavy duty castors
  • Adjustable tray runners and removable recessed top
  • Self assembly

Specifications

Product Type Double Trolley
Color Gray
Description Display: Double trolley with 12 runners
Dimensions (L x W x H) 710 x 420 x 850 mm
Height (Metric) 850 mm
Includes 12 runners
Material Stainless Steel
Size Double
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.