missing translation for 'onlineSavingsMsg'
Learn More

Lithium bromide, ultra dry, 99.998% (metals basis)

Product Code. 11399698
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399698 5 g 5g
11379698 25 g 25g
11389698 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399698 Supplier Thermo Scientific Alfa Aesar Supplier No. 044199.06
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7550-35-8 | BrLi | 86.844 g/mol

Employed as adsorbents, functional fluids (closed systems) and intermediates. Used as an operating medium for air-conditioning and industrial drying system. It is also used in manufacturing pharmaceuticals and alkylation process. It is used as brazing and welding fluxes. Used for reconstitution of brines, as swelling agent for proteins, as electrolyte component in lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7550-35-8
Molecular Formula BrLi
Molecular Weight (g/mol) 86.844
MDL Number MFCD00011077
InChI Key AMXOYNBUYSYVKV-UHFFFAOYSA-M
Synonym lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
PubChem CID 82050
ChEBI CHEBI:63042
IUPAC Name lithium;bromide
SMILES [Li+].[Br-]

Specifications

Boiling Point 1265°C
Melting Point 550°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.998% (Metals basis)
Solubility Information Soluble in water (at 20°C 1490g/L),alcohol and glycol.
Formula Weight 86.85
Sensitivity Hygroscopic
Density 3.464
Chemical Name or Material Lithium bromide, ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.