Learn More
Lithium bromide hydrate, Puratronic™, 99.995% (metals basis)
Description
Lithium bromide is used as a medium for air-conditioning, and a desiccant industrial drying system. It finds use as a sedative and hypnotic. Further it is employed in manufacturing pharmaceuticals, in alkylation processes, and as brazing and welding fluxes. In addition, lithium bromide-water solution is widely used in absorption refrigeration systems, absorption heat pumps and absorption heat transformers. l-Proline lithium bromide monohydrate (LPLBM) is a promising semi-organic nonlinear optical material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 162°C to 167°C |
| Color | White |
| Physical Form | Crystalline Aggregates |
| Quantity | 25 g |
| Assay Percent Range | 99.995% (Metals basis) |
| Solubility Information | Soluble in water,alcohol and glycol. |
| Formula Weight | 86.85 (Anhydrous) |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Chemical Name or Material | Lithium bromide hydrate |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.