missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium bromide, Free-flowing, Reagent Grade, anhydrous, ≥99%, Solstice
Shop All Honeywell Fluka ProductsDescription
- ≥95% for general laboratory use.
Specifications
Specifications
| Boiling Point | 1,265°C |
| Melting Point | 550°C (literature) |
| CAS Max % | 100.0000% |
| Quantity | 100 g |
| Linear Formula | LiBr |
| Formula Weight | 86.85 |
| Percent Purity | ≥99% |
| Packaging | Poly Bottle |
| Density | 3.46 g/cm3 |
| Chemical Name or Material | Lithium bromide, Free-flowing, Reagent Grade, anhydrous, ≥99% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?