missing translation for 'onlineSavingsMsg'
Learn More

Lithium Bromide Crystals, Ultra-Pure Additives, SPEX CertiPrep™

Catalog Number 15276046
Click to view available options
Quantity:
125 g
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Lithium Bromide Crystals, Ultra-Pure Additives, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Ultra Pure Additives
  • Lithium Bromide Crystals
  • 125 g Package Size

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Quantity 125 g
Grade Ultrapure
Packaging Triple-leached LDPE bottle
Chemical Name or Material Lithium Bromide Crystals, Additive Ultra Pure
compliance-icons compliance-icons
Product Identifier
  • FUSION FLUX-ULTRA PURE ADDITIVE
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : lithium bromide

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'