missing translation for 'onlineSavingsMsg'
Learn More

Lithium borohydride, 95%

Product Code. 10422211 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10422211 10 g 10g
10438443 5 g 5g
10308490 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10422211 Supplier Thermo Scientific Acros Supplier No. 206810100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16949-15-8 | BH4Li | 21.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16949-15-8
Molecular Formula BH4Li
Molecular Weight (g/mol) 21.78
MDL Number MFCD00011088
InChI Key UUKMSDRCXNLYOO-UHFFFAOYSA-N
Synonym borate 1-, tetrahydro-, lithium, unii-8l87x4s4kp, boryllithium, lithium, boryl, lithium hydroborate, lithotab borohydride, bh4li, lithium 1+ ion boranuide, lithium borohydride 2.0 m in thf, borate 1-, tetrahydro-, lithium 1:1
PubChem CID 20722760
SMILES [Li+].[BH4-]

Specifications

Melting Point 280.0°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 95%
Linear Formula LiBH4
Fieser 01,603; 04,296; 11,293; 12,276; 13,92; 14,191; 15,186
Merck Index 15, 5581
Solubility Information Solubility in water: 209g/L (10°C). Other solubilities: soluble in ether, ethanol, tetrahydrofuran and, aliphatic amines
IUPAC Name lithium;boron(1-)
Formula Weight 21.77
Percent Purity 95%
Packaging Glass Bottle
Specific Gravity 0.66
Density 0.6600g/mL
Chemical Name or Material Lithium borohydride, 95%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lithium borohydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.