missing translation for 'onlineSavingsMsg'
Learn More

Lithium Standard For ICP, 10,000μg/mL, SPEX CertiPrep™

Product Code. 10603103 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10603103 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10603103 Supplier SPEX Certiprep Supplier No. PLLI13X
  • ONLINE EXCLUSIVE PRICE
    £274.77
    £228.13 / 500mL
    Save £46.64
    16% Off
 Restricted Item
Estimated Shipment: 21-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Li10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
CAS 554-13-2, 7647-01-0, 7732-18-5
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 10.6, 0.1, 84.4
Analyte or Component Names Li
Molecular Formula CLi2O3
MDL Number MFCD00011084
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Type Certified Reference Material
SMILES [Li+].[Li+].[O-]C([O-])=O
Element Li
IUPAC Name dilithium(1+) carbonate
Solvent or Matrix 5% HCl
Molecular Weight (g/mol) 73.89
PubChem CID 11125
Quantity 500 mL
ChEBI CHEBI:6504
Grade Assurance
Concentration or Composition (by Analyte or Components) 1000mg/L
Chemical Name or Material Lithium Standard, For AA and ICP
Show More Show Less
compliance-icons
Product Identifier
  • 10,000 µg/mL Lithium
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: lithium carbinate, hydrochloric acid

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.