missing translation for 'onlineSavingsMsg'
Learn More

Lithium amide, 95%, Thermo Scientific™

Product Code. 10296730 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10296730 500 g Glass bottle 500g
10471051 5 g Glass bottle 5g
10471241 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10296730 Supplier Thermo Scientific Acros Supplier No. 199865000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-89-0
MDL Number MFCD00011093
InChI Key AFRJJFRNGGLMDW-UHFFFAOYSA-N
Synonym lithium amide, lithamide, lithium amide li nh2, lithiumamide, unii-7393omu9lk, lithium azanide, hsdb 649, linh2, lithium amide 25g, lithium amide, powder
PubChem CID 24532
IUPAC Name lithium;azanide
SMILES [Li+].[NH2-]

Specifications

Color White through Gray
Physical Form Fine Powder
Quantity 500 g
Assay Percent Range 95%
Molecular Formula H2LiN
Linear Formula LiNH2
Fieser 01,600; 03,178; 05,392; 13,159; 17,311
Merck Index 15, 5578
Molecular Weight (g/mol) 22.963
Formula Weight 22.95
Percent Purity ≥94% (LiNH2)
Packaging Glass bottle
Density 1.1700g/mL
Chemical Name or Material Lithium amide, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.