missing translation for 'onlineSavingsMsg'
Learn More

Lithium aluminum oxide

Product Code. 11306748
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11306748 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11306748 Supplier Thermo Scientific Alfa Aesar Supplier No. 013412.A1
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 12003-67-7 | AlLiO2 | 65.92 g/mol

It is used as an inert electrolyte support material in molten carbonate fuel cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12003-67-7
Molecular Formula AlLiO2
Molecular Weight (g/mol) 65.92
MDL Number MFCD00016172
InChI Key YQNQTEBHHUSESQ-UHFFFAOYSA-N
Synonym lithium aluminate, lithium metaaluminate, aluminum lithium dioxide, aluminate alo21-, lithium, lithium aluminum spinal, lithium aluminate lialo2, hsa 10, aluminum lithium oxide, aluminum lithium oxide lialo2, lithium aluminum oxide lialo2
PubChem CID 123268
IUPAC Name lithium;oxido(oxo)alumane
SMILES [Li+].[O-][Al]=O

Specifications

Melting Point >1625°C
Color White
Physical Form Powder
Quantity 1 kg
Odor Odorless
Density 2.62
Solubility Information Insoluble in water.
Formula Weight 65.92
Chemical Name or Material Lithium aluminum oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.