missing translation for 'onlineSavingsMsg'
Learn More

Lithium aluminum hydride, 95%, powder

Product Code. 10042470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
5 kg
Unit Size:
100g
10g
25g
500g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10042470 10 g 10g
10665832 25 g 25g
10123830 100 g 100g
10207003 500 g 500g
17332481 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10042470 Supplier Thermo Scientific Acros Supplier No. 190320100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 16853-85-3 | AlH4Li | 37.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16853-85-3
Molecular Formula AlH4Li
MDL Number MFCD00011075
Synonym lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
PubChem CID 21226445

Specifications

Melting Point 125.0°C
Physical Form Powder
Quantity 10 g
Assay Percent Range 95%
Linear Formula LiAlH4
Fieser 01,581; 02,242; 03,176; 04,291; 05,382; 06,325; 07,196; 08,286; 09,274; 10,236; 11,289; 12,272; 13,61; 14,190; 15,184; 16,133; 17,162
Merck Index 15, 344
Solubility Information Solubility in water: reacts. Other solubilities: soluble in diethyl ether (39g/100g at 25 °C), thf/, toluene (70:30) (17g/100g at 25 °C), in thf/mtbe (, 25:75) (12g/100g at 25 °C), in thf/toluene (15:85), (5g/100g at 25 °C);, practically insoluble in hyd
InChI Key OCZDCIYGECBNKL-UHFFFAOYSA-N
SMILES [Li+].[AlH4-]
IUPAC Name lithium(1+) alumanuide
Molecular Weight (g/mol) 37.95
Formula Weight 37.95
Percent Purity 95%
Density .9g/mL
Chemical Name or Material Lithium Aluminum hydride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium aluminium hydride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.