missing translation for 'onlineSavingsMsg'
Learn More

Lithium Acetate Dihydrate, Technical, SLR, Fisher Chemical

Product Code. 10355900 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic powder jar
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10355900 250 g Plastic powder jar 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10355900 Supplier Fisher Chemical Supplier No. L/2050/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 6108-17-4
Melting Point 56°C
Color White
pH 7.5 to 9.5
Packaging Plastic powder jar
Molecular Formula C2H7LiO4
MDL Number 66949
Quantity 250 g
Synonym lithium acetate dihydrate, lithiumacetatedihydrate, lithium acetate dihydrate, reagent grade, lioac-2h2o, lioac.2h2o, ksc498a1d, c2h3o2.li.2h2o, lithotab acetate ion dihydrate, acetic acid lithium salt dihydrate
InChI Key IAQLJCYTGRMXMA-UHFFFAOYSA-M
SMILES [Li+].CC(=O)[O-].O.O
IUPAC Name lithium;acetate;dihydrate
Molecular Weight (g/mol) 102.014
PubChem CID 23666338
Formula Weight 102.02g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.