missing translation for 'onlineSavingsMsg'
Learn More

Lithium acetate dihydrate, 98%, extra pure

Product Code. 10450861 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10450861 1 kg Plastic bottle 1kg
10730901 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10450861 Supplier Thermo Scientific Acros Supplier No. 199840010
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6108-17-4 | C2H3LiO2·2H2O | 102.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6108-17-4
Molecular Formula C2H3LiO2·2H2O
Molecular Weight (g/mol) 102.02
MDL Number MFCD00066949
InChI Key IAQLJCYTGRMXMA-UHFFFAOYSA-M
Synonym lithium acetate dihydrate, lithiumacetatedihydrate, lithium acetate dihydrate, reagent grade, lioac-2h2o, lioac.2h2o, ksc498a1d, c2h3o2.li.2h2o, lithotab acetate ion dihydrate, acetic acid lithium salt dihydrate
PubChem CID 23666338
IUPAC Name lithium;acetate;dihydrate
SMILES [Li+].CC(=O)[O-].O.O

Specifications

Melting Point 56°C to 58°C
Color White
pH 7.5 to 9.5 (5% soln.)
Physical Form Adhering Crystalline Powder and Chunks
Quantity 1 kg
Assay Percent Range 98%
Linear Formula CH3CO2Li·2H2O
Beilstein 02, 107
Merck Index 15, 5577
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: very soluble in hot water,215g/L ethanol
Formula Weight 102.02
Percent Purity 98%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Lithium acetate dihydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.