missing translation for 'onlineSavingsMsg'
Learn More

Lithium-6 Internal Standard: 6Li @ 100 μg/mL in 2% HNO3, VHG™

Product Code. 17998809
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17998809 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17998809 Supplier VHG Labs Supplier No. VHGLISC6LI100

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Lithium-6 Internal Standard: 6Li @ 100 μg/mL in 2% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium
CAS 7439-93-2
Methods ICP-MS
Analyte or Component Names Lithium
Shipping Condition Room Temperature
Molecular Formula Li
Certifications/Compliance ISO 17034
Synonym Li
InChI Formula InChI=1S/Li
SMILES [Li]
IUPAC Name Lithium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 6.94
Quantity 100 mL
Formula Weight 7.016 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.