missing translation for 'onlineSavingsMsg'
Learn More

Lithium, 1000μg/mL (0.10% w/v), BAKER INSTRA-ANALYZED™ Reagent, J.T. Baker™

Product Code. 15577644 Shop All J.T.Baker Products
Click to view available options
Quantity:
150 mL
Unit Size:
150mL
This item is not returnable. View return policy

Product Code. 15577644

Brand: Avantor J.T.Baker 645604

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

Specifications

Packaging Narrow Mouth Poly Bottle
Specific Gravity 1L = 1.01kg
Molecular Formula Li
Quantity 150 mL
Formula Weight 6.94
Grade BAKER INSTRA-ANALYZED™ Reagent
Chemical Name or Material Lithium, 1000μg/mL (0.10% w/v)
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362-Take off contaminated clothing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.