missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lissamine rhodamine B sulfonyl chloride, pure

Product Code. 10536792 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10536792 5 g Glass bottle 5g
10167940 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10536792 Supplier Thermo Scientific Chemicals Supplier No. 413230050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62796-29-6 | C27H29ClN2O6S2 | 577.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62796-29-6
Molecular Formula C27H29ClN2O6S2
Molecular Weight (g/mol) 577.107
MDL Number MFCD00042001
InChI Key YERWMQJEYUIJBO-UHFFFAOYSA-N
Synonym Sulforhodamine B sulfonyl chloride
PubChem CID 65223
IUPAC Name 5-chlorosulfonyl-2-[3-(diethylamino)-6-diethylazaniumylidenexanthen-9-yl]benzenesulfonate
SMILES CCN(CC)C1=CC2=C(C=C1)C(=C3C=CC(=[N+](CC)CC)C=C3O2)C4=C(C=C(C=C4)S(=O)(=O)Cl)S(=O)(=O)[O-]

Specifications

Assay Percent Range 99+%
Infrared Spectrum Authentic
Formula Weight 577.11
Grade Pure
Packaging Glass bottle
UV Spectrometry Molar ext.%gt; 85, 000L/mol.cm(lambda 564 +/- 3nm) in dichloromethane
Color Green
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Lissamine rhodamine B sulfonyl chloride, 99+%
compliance-icons
Product Identifier
  • Lissamine rhodamine B sulfonyl chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.