missing translation for 'onlineSavingsMsg'
Learn More

LIS1 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18420924 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420924 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420924 Supplier Novus Biologicals Supplier No. NBP176815IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LIS1 Polyclonal antibody specifically detects LIS1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIS1
Applications Western Blot, ELISA, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias LIS-1, LIS1PAF-AH alpha, LIS2, lissencephaly 1 protein, Lissencephaly-1 protein, MDCR, MDS, Miller-Dieker syndrome chromosome region, PAF acetylhydrolase 45 kDa subunit, PAFAH, PAF-AH 45 kDa subunit, PAFAH alpha, PAFAHA, platelet-activating factor acetylhydrolase 1b, regulatory subunit 1 (45kDa), platelet-activating factor acetylhydrolase IB subunit alpha, platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit (45kD), platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit 45kDa, platelet-activating factor acetylhydrolase, isoform Ib, subunit 1 (45kDa)
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human LIS1. The immunogen is located within amino acids 340 - 390 of LIS1. Amino Acid Squence: DDKTLRVWDYKNKR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5048
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.