missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lirilumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558346
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Lirilumab, also known as IPH2102 or BMS-986015, is a fully human IgG4 monoclonal antibody targeting inhibitory killer-cell immunoglobulin-like receptors (KIRs), notably KIR2DL1, KIR2DL2, and KIR2DL3, as well as activating KIR2DS1 and KIR2DS2. The structure of lirilumab includes the variable domains which bind to a conformational epitope on domain 1 of KIR2DL3. This binding does not significantly alter the structure of KIR2DL3 in comparison to its unbound form. Lirilumab's binding epitope overlaps with the HLA binding site, which influences the clinical efficacy of the drug as variations in this epitope among patients may impact its effectiveness. Functionally, lirilumab blocks inhibitory KIRs, thereby enhancing NK cell activity against tumor cells. This mechanism underlies its investigation in clinical trials for hematologic and solid malignancies, evaluating its efficacy both as a monotherapy and in combination with other agents such as azacitidine.
Specifications
| Lirilumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| BMS-986015; IPH2102 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG4SP |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction