missing translation for 'onlineSavingsMsg'
Learn More

Liranaftate, MedChemExpress

Product Code. 16444337
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16444337

Brand: Medchem Express HYB0348

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Liranaftate (Piritetrate) is a squalene epoxidase inhibitor with anti-fungicidal activities. Liranaftate can be used for the research of dermatophytes. Liranaftate also suppresses fungal element-promoted production of IL-8 and experimental inflammation.

Liranaftate (Piritetrate) is a squalene epoxidase inhibitor with anti-fungicidal activities. Liranaftate can be used for the research of dermatophytes. Liranaftate also suppresses fungal element-promoted production of IL-8 and experimental inflammation.

Specifications

Chemical Name or Material Liranaftate
Physical Form Solid
CAS 88678-31-3
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H20N2O2S
For Use With (Application) COVID-19-immunoregulation
Synonym Piritetrate M-732
Solubility Information DMSO : 100 mg/mL (304.48 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES S=C(OC1=CC=C2CCCCC2=C1)N(C3=NC(OC)=CC=C3)C
Molecular Weight (g/mol) 328.43
Formula Weight 328.43
Percent Purity 99.4%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.