Learn More
LIR-8/CD85c/LILRB5 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98702MFV610
This item is not returnable.
View return policy
Description
LIR-8/CD85c/LILRB5 Polyclonal antibody specifically detects LIR-8/CD85c/LILRB5 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| LIR-8/CD85c/LILRB5 | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Store at 4C in the dark. |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| 10990 |