missing translation for 'onlineSavingsMsg'
Learn More

LIR-8/CD85c/LILRB5 Antibody (1058033) [DyLight 405], Novus Biologicals™

Product Code. 30737395 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737395 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737395 Supplier Novus Biologicals Supplier No. FAB30651E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LIR-8/CD85c/LILRB5 Monoclonal antibody specifically detects LIR-8/CD85c/LILRB5 in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIR-8/CD85c/LILRB5
Applications Immunocytochemistry
Classification Monoclonal
Clone 1058033
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CD85 antigen-like family member C, CD85c, CD85c antigen, Leukocyte immunoglobulin-like receptor 8, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR8CD85C, LIR-8leukocyte immunoglobulin-like receptor subfamily B member 5, member 5
Host Species Mouse
Immunogen Mouse myeloma cell line, NS0-derived human LILRB5/CD85c/LIR-8, Arg18-His456, Accession # O75023
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10990
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.