missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ LIR-6/LILRA1 Antibody (4A1E10A6), Novus Biologicals™

Mouse Monoclonal Antibody

£236.00 - £406.00

Specifications

Antigen LIR-6/LILRA1
Clone 4A1E10A6
Concentration 1 mg/mL
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000
Applications Flow Cytometry, ELISA, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30028594
View Documents
Novus Biologicals™
NBP3-27141-0.1ml
0.1 mL
£406.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30038208
View Documents
Novus Biologicals™
NBP3-27141-0.025ml
0.025 mL
£236.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LIR-6/LILRA1 Monoclonal antibody specifically detects LIR-6/LILRA1 in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot
Specifications

Specifications

LIR-6/LILRA1
1 mg/mL
Flow Cytometry, ELISA, Western Blot
Unconjugated
Mouse
Human
CD85 antigen-like family member I, CD85i, CD85i antigen, leukocyte immunoglobulin-like receptor subfamily A member 1, leukocyte immunoglobulin-like receptor subfamily A member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 1, LIR-6Leukocyte immunoglobulin-like receptor 6, LIR6MGC126563
Purified recombinant fragment of human LIR-6/LILRA1 (AA: extra 322-461) expressed in E. Coli.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
4A1E10A6
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000
Monoclonal
Purified
RUO
PBS
11024
IgG2a
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.