missing translation for 'onlineSavingsMsg'
Learn More

LIR-6/LILRA1 Antibody (356316) [DyLight 405], Novus Biologicals™

Product Code. 30731690 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30731690 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731690 Supplier Novus Biologicals Supplier No. FAB3085E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LIR-6/LILRA1 Monoclonal antibody specifically detects LIR-6/LILRA1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIR-6/LILRA1
Applications Western Blot
Classification Monoclonal
Clone 356316
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CD85 antigen-like family member I, CD85i, CD85i antigen, leukocyte immunoglobulin-like receptor subfamily A member 1, leukocyte immunoglobulin-like receptor subfamily A member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 1, LIR-6Leukocyte immunoglobulin-like receptor 6, LIR6MGC126563
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LILRA1/CD85i/LIR-6, Pro17-Asn461, Accession # O75019
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11024
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.