missing translation for 'onlineSavingsMsg'
Learn More

Azpack Limited™ Liqui-Nox Precision Liquid Detergent

Product Code. 12777137
This item is not returnable. View return policy

Product Code. 12777137

Brand: Azpack Limited™ 12777137

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rinses easily under runinng water without leaving residue. Azpack Limited™ Liqui-Nox Precision Liquid Detergent is extremely mild and completely soluble in hard and soft water.

  • Phosphate free, biodegradable, and exceptionally mild to skin.
  • Not low-foamoing: Do not use in automated cleaners or processors.
  • Contains no ammonia.
  • Contains EDTA “ethylene diamine tetra acetate” which will not be detected nor interfere with a Kjeldahl nitrogen determination.
  • Does not support bacterial growth in concentrated form. When diluted, bacteria may grow and produce ammonia when they eat the EDTA. In these cases, a positive reading on total nitrogen could be indicative to the presence of nitrogen.
  • Shelf life: one year, from the date of shipment, which corresponds to the date for which the performance is guaranteed by the manufacturer.
  • Guaranteed in terms of its performance for two years after manufacturing, whichever date comes first.
  • Dilute 1:100.
  • Final solution will have a pH of 8.5
  • Packaged in plastic bottles

Specifications

Product Type Liquid Detergent
pH 8.5 (1%)
Certifications/Compliance USDA authorized
Composition EDTA
Quantity 1 Ea.
Volume (English) 1 gal.
compliance-icons
Product Identifier
  • Liqui-nox phosphate-free liquid detergent
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P264-Wash skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : benzene sulphonic acid, sodium xylenesulphonate, C12-C14 secondary alcohols, coconut diethanolamide,
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.