missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Liq-Glass

£159.00 - £162.00

Specifications

For Use With (Application) For daily laboratory use, titrations with strong acids as well as in strong bases
Temperature (Metric) Operating -10°C to +100°C
Length (Metric) 120 mm
Type Liq-Glass
pH Range 0 to 14
View More Specs

Products 2
Product Code Brand Connector Types Price Quantity & Availability  
Product Code Brand Connector Types Price Quantity & Availability  
10083543
View Documents Promotion Details
Hamilton™
238185
DIN
£162.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 28-06-2022
to see stock.
Add to basket
 
11590162
View Documents Promotion Details
Hamilton™
238180
BNC
£159.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 28-06-2022
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

For daily laboratory use, titrations with strong acids as well as in strong bases
120 mm
0 to 14
-10°C to +100°C
Liq-Glass
SDS
Product Certifications