missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Liq-Glass

Product Code. 10083543
Change view
Click to view available options
Connector Types:
BNC
DIN
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types unitSize
10083543 DIN Each
11590162 BNC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10083543 Supplier Hamilton™ Supplier No. 238185

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Connector Types DIN
Type Liq-Glass
For Use With (Application) For daily laboratory use, titrations with strong acids as well as in strong bases
pH Range 0 to 14
Temperature (Metric) Operating -10°C to +100°C
Length (Metric) 120 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.