missing translation for 'onlineSavingsMsg'
Learn More

Liproxstatin-1, MedChemExpress

Product Code. 16485956
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485956 100 mg 100mg
16481507 50 mg 50mg
16458017 5 mg 5mg
16415308 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485956 Supplier Medchem Express Supplier No. HY12726
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Liproxstatin-1 is a potent ferroptosis inhibitor and inhibits ferroptotic cell death (IC50=22 nM).

Liproxstatin-1 is a potent ferroptosis inhibitor and inhibits ferroptotic cell death (IC50=22 nM).

Specifications

Chemical Name or Material Liproxstatin-1
CAS 950455-15-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H21ClN4
Quantity 100 mg
Solubility Information DMSO : ≥ 31 mg/mL (90.95 mM)
SMILES ClC1=CC(CNC2=NC3=CC=CC=C3NC24CCNCC4)=CC=C1
Molecular Weight (g/mol) 340.85
Formula Weight 340.85
Percent Purity 97.13%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.