missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liprin alpha 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14175-1-AP-150UL
This item is not returnable.
View return policy
Description
May regulate the disassembly of focal adhesions. May localize receptor-like tyrosine phosphatases type 2A at specific sites on the plasma membrane.Specifications
| Liprin alpha 1 | |
| Polyclonal | |
| Unconjugated | |
| PPFIA1 | |
| LAR interacting protein 1, LIP 1, LIP.1, LIP1, LIPRIN, Liprin alpha 1, PPFIA1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 233977, 293645, 8500 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13136 | |
| PPFIA1 | |
| Liprin alpha 1 Fusion Protein Ag5381 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction