missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liprin alpha 1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £375.00
Specifications
| Antigen | Liprin alpha 1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18361486
|
Novus Biologicals
NBP3-03786-20UL |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18308486
|
Novus Biologicals
NBP3-03786-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Liprin alpha 1 Polyclonal specifically detects Liprin alpha 1 in Human, Mouse samples. It is validated for Western Blot.Specifications
| Liprin alpha 1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 8500 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| FLJ42630, FLJ43474, LAR-interacting protein 1, LIP.1, LIP-1, LIP1FLJ41337, LIPRIN, liprin-alpha-1, Liprin-alpha1, MGC26800, Protein tyrosine phosphatase receptor type f polypeptide-interacting proteinalpha-1, protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interactingprotein (liprin), alpha 1, PTPRF-interacting protein alpha-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 660-800 of human Liprin alpha 1 (NP_003617.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title