missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liprin alpha 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £470.00
Specifications
| Antigen | Liprin alpha 1 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18245661
|
Novus Biologicals
NBP2-58915 |
100 μL |
£470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663338
|
Novus Biologicals
NBP2-58915-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
Liprin alpha 1 Polyclonal specifically detects Liprin alpha 1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Tekniske data
| Liprin alpha 1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 8500 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:HHKALDEKVRERLRVALERCSLLEEELGATHKELMILKEQNNQKKTLTDGVLDINHEQENTPSTSGKRSSDGSLSHEEDLAKVIELQEIISKQSREQSQMKERLASLSSHVTELEEDLDTAR | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| FLJ42630, FLJ43474, LAR-interacting protein 1, LIP.1, LIP-1, LIP1FLJ41337, LIPRIN, liprin-alpha-1, Liprin-alpha1, MGC26800, Protein tyrosine phosphatase receptor type f polypeptide-interacting proteinalpha-1, protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interactingprotein (liprin), alpha 1, PTPRF-interacting protein alpha-1 | |
| PPFIA1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel