missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liprin alpha 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | Liprin alpha 1 |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18114048
|
Novus Biologicals
NBP2-38494 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18607215
|
Novus Biologicals
NBP2-38494-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Liprin alpha 1 Polyclonal specifically detects Liprin alpha 1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Liprin alpha 1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q13136 | |
| 8500 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: QLEETQHDKDQLVLNIEALRAELDHMRLRGASLHHGRPHLGSVPDFRFPMADGHTDSYSTSAVLRRPQKGRLAALRDEPSKVQTLNEQDWERAQQASVLANVAQAFESDADVSDGEDDRDTLLSSVDLLS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FLJ42630, FLJ43474, LAR-interacting protein 1, LIP.1, LIP-1, LIP1FLJ41337, LIPRIN, liprin-alpha-1, Liprin-alpha1, MGC26800, Protein tyrosine phosphatase receptor type f polypeptide-interacting proteinalpha-1, protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interactingprotein (liprin), alpha 1, PTPRF-interacting protein alpha-1 | |
| PPFIA1 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title