missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liprin alpha 1 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
£245.00
Specifications
| Antigen | Liprin alpha 1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Chicken |
Description
Liprin alpha 1 Polyclonal specifically detects Liprin alpha 1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| Liprin alpha 1 | |
| Polyclonal | |
| Chicken | |
| Human | |
| Q13136 | |
| 8500 | |
| The immunogen for this antibody was a sequence from 310a.a. to 360a.a. from Liprin-alpha-1(a 1202a.a. long protein) | |
| Affinity Purified | |
| Liprin-alpha-1 |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with No Preservative | |
| FLJ42630, FLJ43474, LAR-interacting protein 1, LIP.1, LIP-1, LIP1FLJ41337, LIPRIN, liprin-alpha-1, Liprin-alpha1, MGC26800, Protein tyrosine phosphatase receptor type f polypeptide-interacting proteinalpha-1, protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interactingprotein (liprin), alpha 1, PTPRF-interacting protein alpha-1 | |
| PPFIA1 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title