missing translation for 'onlineSavingsMsg'
Learn More

Lipoprotein Lipase/LPL Antibody (OTI3A10), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-71178PCP

Product Code. 18188763

  • £639.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Lipoprotein Lipase/LPL Monoclonal specifically detects Lipoprotein Lipase/LPL in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lipoprotein Lipase/LPL
Monoclonal
PerCP
PBS with 0.05% Sodium Azide
LPL
Recombinant protein fragment corresponding to amino acids 28-475 of human Lipoprotein Lipase/LPL (NP_000228) produced in HEK293T.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Flow Cytometry
OTI3A10
Flow Cytometry
EC 3.1.1, HDLCQ11, LIPDEC 3.1.1.34, lipoprotein lipase
Mouse
Affinity Purified
Lipid and Metabolism
4023.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.