missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipoprotein Lipase/LPL Antibody (OTI3A10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-01395
This item is not returnable.
View return policy
Description
Lipoprotein Lipase/LPL Monoclonal specifically detects Lipoprotein Lipase/LPL in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Lipoprotein Lipase/LPL | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Flow Cytometry 1:100, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
| LPL | |
| Human recombinant protein fragment corresponding to amino acids 28-475 of human LPL(NP_000228) produced in HEK293T. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI3A10 | |
| Unconjugated | |
| EC 3.1.1, HDLCQ11, LIPDEC 3.1.1.34, lipoprotein lipase | |
| Mouse | |
| 50.3 kDa | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 4023 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction