missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipoprotein Lipase/LPL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32215
This item is not returnable.
View return policy
Description
Lipoprotein Lipase/LPL Polyclonal specifically detects Lipoprotein Lipase/LPL in Human, Mouse, Chicken, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Lipoprotein Lipase/LPL | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| LPL | |
| Recombinant protein encompassing a sequence within the center region of human LPL. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.59mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| EC 3.1.1, HDLCQ11, LIPDEC 3.1.1.34, lipoprotein lipase | |
| Rabbit | |
| 53 kDa | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 4023 | |
| Human, Mouse, Chicken | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction