missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipoprotein Lipase/LPL Antibody (2C5), Novus Biologicals™
Mouse Monoclonal Antibody
£161.00
Specifications
| Antigen | Lipoprotein Lipase/LPL |
|---|---|
| Clone | 2C5 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Lipoprotein Lipase/LPL Monoclonal specifically detects Lipoprotein Lipase/LPL in Human samples. It is validated for Western Blot, ELISA.Specifications
| Lipoprotein Lipase/LPL | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 4023 | |
| Purified recombinant fragment of LPL expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 2C5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism | |
| EC 3.1.1, HDLCQ11, LIPDEC 3.1.1.34, lipoprotein lipase | |
| LPL | |
| IgG1 | |
| Unpurified | |
| 53.1 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title