missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Lipopolysaccharide (LPS) Solution (500X)

Product Code. 15536286
Change view
Click to view available options
Quantity:
100 μL
4 x 100 μL
Unit Size:
100µL
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15536286 100 μL 100µL
15526286 4 x 100 μL Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15536286 Supplier Invitrogen™ Supplier No. 00497693

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Intended for in vitro culture of B cells, monocytes, macrophages, and other cells that are responsive to LPS.

  • A ready to use solution of LPS from Escherichia coli 026:B6 in aqueous solution
  • LPS is a major component of the cell wall of gram negative bacteria
  • It is highly immunogenic and strongly activates immune cells bearing the CD14/TLR4/MD2 receptor complex
  • This reagent is intended for use in in vitro culture of B cells, monocytes, macrophages and other cells that are responsive to LPS
TRUSTED_SUSTAINABILITY

Specifications

Cell Type B cells, monocytes, macrophages and other cells that are responsive to LPS.
Concentration 500 X
Form Liquid
Product Type LPS Solution
Sterility Sterile
Product Line eBioscience
Quantity 100 μL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.