Learn More
Invitrogen™ eBioscience™ Lipopolysaccharide (LPS) Solution (500X)
Intended for in vitro culture of B cells, monocytes, macrophages, and other cells that are responsive to LPS.
Brand: Invitrogen™ 00-4976-03
113.28 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.09500kg
Description
- A ready to use solution of LPS from Escherichia coli 026:B6 in aqueous solution
- LPS is a major component of the cell wall of gram negative bacteria
- It is highly immunogenic and strongly activates immune cells bearing the CD14/TLR4/MD2 receptor complex
- This reagent is intended for use in in vitro culture of B cells, monocytes, macrophages and other cells that are responsive to LPS
Specifications
B cells, monocytes, macrophages and other cells that are responsive to LPS. | |
Liquid | |
Sterile | |
4 x 100 μL |
500 X | |
LPS Solution | |
eBioscience™ |
For Research Use Only. Not for use in diagnostic procedures.