missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipocalin-2/NGAL Rabbit anti-Human, Clone: 128, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | Lipocalin-2/NGAL |
|---|---|
| Clone | 128 |
| Dilution | Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18211306
|
Novus Biologicals
NBP2-89346-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18233478
|
Novus Biologicals
NBP2-89346-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Human samples. It is validated for Western Blot, ELISA.Specifications
| Lipocalin-2/NGAL | |
| Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin | |
| Recombinant Human Lipocalin-2/NGAL protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 128 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuroscience | |
| 0.2 um filtered solution in PBS | |
| 3934 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title