missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipocalin-2/NGAL Mouse anti-Porcine, Clone: 17, Novus Biologicals™
Mouse Monoclonal Antibody
£512.00
Specifications
| Antigen | Lipocalin-2/NGAL |
|---|---|
| Clone | 17 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:8000 |
| Applications | Western Blot |
Description
Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Porcine samples. It is validated for Western Blot, ELISA.Specifications
| Lipocalin-2/NGAL | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl | |
| 3934 | |
| IgG1 κ | |
| Protein A or G purified |
| 17 | |
| Western Blot 1:100-1:2000, ELISA 1:8000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Pig | |
| 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin | |
| Recombinant pig NGAL adsorbed onto aluminum hydroxide gel | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title