missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipocalin-2/NGAL Mouse anti-Canine, Clone: 14, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-41364
This item is not returnable.
View return policy
Description
Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Canine, Human (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Sandwich ELISA, Sandwich ELISA Detection.
Specifications
| Lipocalin-2/NGAL | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Sandwich ELISA, Sandwich ELISA Capture | |
| 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin | |
| Recombinant dog NGAL adsorbed onto aluminum hydroxide gel | |
| 100 μg | |
| Primary | |
| Canine, Human (Negative), Mouse (Negative), Rat (Negative) | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| 14 | |
| Unconjugated | |
| 10 mM Phosphate (pH 7.4) and 0.5 M NaCl | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 3934 | |
| Store at 4°C. Do not freeze. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction