missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipocalin-2/NGAL Antibody (972737), Alexa Fluor 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB17572AF488
This item is not returnable.
View return policy
Description
Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Lipocalin-2/NGAL | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LCN2 | |
| Mouse myeloma cell line NS0-derived recombinant human Lipocalin-2/NGAL | |
| 100 μL | |
| Lipid and Metabolism | |
| 3934 | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 972737 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction