missing translation for 'onlineSavingsMsg'
Learn More

Lipocalin-2/NGAL Antibody (128), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18369810 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369810 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369810 Supplier Novus Biologicals Supplier No. NBP289346AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lipocalin-2/NGAL
Applications Western Blot, ELISA
Classification Monoclonal
Clone 128
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Lipocalin-2/NGAL (NP_005555.2; Met 1-Gly 198).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3934
Test Specificity No cross-reactivity in ELISA with: Human MMP9 Rat LCN2
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.