missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIPJ Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14195
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
LIPJ Polyclonal specifically detects LIPJ in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Spécification
| LIPJ | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| bA425M17.2, FLJ11218, lipase J, lipase member J, lipase, family member J, lipase-like abhydrolase domain-containing protein 1, lipase-like, ab-hydrolase domain containing 1, LIPL1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 142910 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| LIPJ | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: YNMTNMNVATAIWNGKSDLLADPEDVNILHSEITNHIYYKTISYYNHIDSLFGLDVYDQVYHEIIDIIQD | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu