missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipin 2 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-12783AF350
This item is not returnable.
View return policy
Description
Lipin 2 Polyclonal antibody specifically detects Lipin 2 in Human samples. It is validated for Western Blot
Specifications
| Lipin 2 | |
| Polyclonal | |
| Western Blot | |
| EC 3.1.3.4, KIAA0249lipin-2, lipin 2, Lipin-2, phosphatidate phosphatase LPIN2 | |
| The immunogen this antibody was made to, maps to a region between residue 275 and 325 of human Lipin 2 using the numbering given in entry NP_055461.1 (GeneID 9663). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 9663 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction