missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lipid A Monoclonal Antibody (43)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA140132
This item is not returnable.
View return policy
Description
This antibody reacts with the hydrophilic backbone of free lipid A; it only reacts with lipid A not covalently bound to LPS.
LPS is chemically composed of a lipid part, which is responsible for the endotoxic activities of LPS, and a heteropolysaccharide which generally consists of an oligosaccharide, termed the core region, and, in some bacterial families, of the so-called O chain, composed of repeating oligosaccharide units. Lipid A is the glucosamine-based phospholipid which forms part of the bacterial outer membrane and anchors LPS to the surface of the bacteria.
Specifications
| Lipid A | |
| Monoclonal | |
| 200 μg/mL | |
| Tissue culture supernatant with 0.02% sodium azide | |
| Mouse | |
| 1 mL | |
| Primary | |
| 4°C | |
| Liquid |
| ELISA, Western Blot | |
| 43 | |
| Unconjugated | |
| Lipid A free | |
| Lipid A. | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction