Learn More
Invitrogen™ Lipid A LPS Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA173178
Description
PA1-73178 is specific for Lipid A LPS. It is cross-reactive with numerous members of Enterobacteriaciae. This antibody is a purified IgG fraction; purity level >95%. Store at 2-8°C up to 6 months. For long term storage, aliquot and store at -20°C, avoiding freeze/thaw cycles.
LPS is chemically composed of a lipid part, which is responsible for the endotoxic activities of LPS, and a heteropolysaccharide which generally consists of an oligosaccharide, termed the core region, and, in some bacterial families, of the so-called O chain, composed of repeating oligosaccharide units. Lipid A is the glucosamine-based phospholipid which forms part of the bacterial outer membrane and anchors LPS to the surface of the bacteria.
Specifications
| Lipid A LPS | |
| Polyclonal | |
| Unconjugated | |
| Lipid A Lipopolysaccharide | |
| Whole cell prep of Lipid A from E. coli O157. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry, Western Blot | |
| 5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Goat | |
| IgG fraction | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.