missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIPI Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24300-1-AP-150UL
This item is not returnable.
View return policy
Description
Hydrolyzes specifically phosphatidic acid (PA) to produce 2-acyl lysophosphatidic acid (LPA; a potent bioactive lipid mediator) and fatty acid. Does not hydrolyze other phospholipids, like phosphatidylserine (PS), phosphatidylcholine (PC) and phosphatidylethanolamine (PE) or triacylglycerol (TG).Specifications
| LIPI | |
| Polyclonal | |
| Unconjugated | |
| LIPI | |
| Cancer/testis antigen 17, CT17, Lipase member I, lipase, member I, LIPI, LPD lipase, LPDL, mPA PLA1 beta, PRED5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 149998, 320355 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6XZB0 | |
| Lipi | |
| LIPI Fusion Protein Ag19024 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction